missing translation for 'onlineSavingsMsg'
Learn More

CD11c Mouse anti-Human, V450, Clone: B-LY6, BD

Mouse Monoclonal Antibody

£396.00 - £545.00

Specifications

Antigen CD11c
Clone B-LY6
Applications Flow Cytometry
Classification Monoclonal
Conjugate Violet Fluor 450 (V450)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15890139
View Documents Promotion Details
BD Biosciences
560370
30 Tests
£396.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15880139
View Documents Promotion Details
BD Biosciences
560369
120 Tests
£545.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reacts with the 150kDa adhesion glycoprotein (p150, integrin α chain) expressed on NK cells, a subset of B and T cells, monocytes, granulocytes and macrophages. It associates with CD18 to form the CD11c/CD18 complex, which binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 may play a role as an adhesion molecule which binds to receptors on stimulated epithelium. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

Host Species: Mouse
Clone: B-ly6
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Specifications

CD11c
Flow Cytometry
Violet Fluor 450 (V450)
RUO
Human
Spleen Cells from Human with Hairy Cell Leukemia
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
B-LY6
Monoclonal
Mouse
Immunology
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
ITGAX; AlphaX integrin chain; Axb2; Integrin alpha-X; CR4; Leu M5; SLEB6
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.