missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Anti-Clumping Agent

Anti-Clumping Agent
£56.50 - £312.00
Specifications
Content And Storage | Store at 2 to 8°C |
---|---|
Cell Line | CHO, Insect Cells, 293 (HEK) |
Cell Type | Mammalian Cells |
Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
Culture Type | Suspension Cell Culture |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11574536
|
Gibco™
0010057AE |
20 mL |
£56.50
20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15234475
![]() |
Gibco™
0010057DG |
100 mL |
£312.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GIBCO™ Anti-Clumping Agent is a concentrated liquid reagent that reduces cell clumping to assist in attaining higher viable cell densities in suspension culture. Anti-Clumping Agent is a gentle, animal origin-free, chemically defined formulation that contains no proteins, enzymes, hydrolysates, or components of unknown composition.Simple and Gentle Cell Dissociation
Cell lines adapted to suspension culture can revert to a more adherent or aggregated “clumping” state. When this occurs, culture condition can be greatly improved by adding a gentle reagent to reduce clumping and adhesion instead of the harsh enzymes often found in cell dissociation products. Anti-Clumping Agent is a simple, gentle, concentrated solution which, when added to culture media (usually titrated between a 1:100 and 1:1000 dilution), can reduce cell clumping significantly.
Reduced Clumping, Increased Viability, Improved Protein Expression
Reduced cell clumping achieved through use of Anti-Clumping Agent is illustrated below (Fig 1). See also the data demonstrating improved viable cell density (Fig 2). Higher viable cell density contributes to the stabilization of culture conditions for better protein yield.
For actual cell dissociation, we recommend our animal origin-free TrypLE™ products.
Use Anti-Clumping Agent after Transfection
Anti-Clumping Agent is not recommended in all suspension cell culture conditions. For example, when creating a cell line using lipid cationic transfection, Anti-Clumping Agent can interfere with the transfection process, so in this case we recommend that Anti-Clumping Agent be used after transfection.
Animal Origin Free and Chemically Defined Components
Because Anti-Clumping Agent is made with only animal origin free and chemically defined components, you can be assured that the risk of adding adventitious agents to your culture system is minimized. Avoiding additives in your system that are not well defined can lead to more robust results from batch to batch.
Convenient and Customizable Packaging
In addition to this 20 ml size, Anti-Clumping Agent is also available in a more economical and convenient 100 mL bottle. For even larger packaging, you can custom configure this product by clicking the 'Customize this Product' link under the 'Add to Cart' button above or by contacting us at CustomMedia@lifetech.com.
Additional Information and Support
Please review the Manuals and Protocols provided below for additional information on successful use of this product. Direct any unanswered questions to our Technical Services group.
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Store at 2 to 8°C | |
Mammalian Cells | |
Suspension Cell Culture | |
Anti-clumping Reagent | |
6 to 8 | |
Room Temperature |
CHO, Insect Cells, 293 (HEK) | |
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Liquid | |
Bottle(s) | |
Gibco™ | |
Animal origin-free |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Anti-Clumping Agent