Learn More
GFAP Mouse, Unlabeled, Clone: 4A11, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 556327
309.19 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
GFAP (Glial Fibrillary Acid Protein) is the major protein of glial filaments in differentiated astrocytes. BD Biosciences Pharmingen offers a panel of monoclonal antibodies (4A11, 1B4, 2E1) that specifically recognize GFAP. They do not cross-react with other intermediate filaments such as vimentin, neurofilament proteins, desmin, keratin, neurotubules or microfilaments. Clone 4A11 is specific for GFAP. Bovine spinal cord homogenate was used as immunogen. This antibody has broad species reactivity, recognizing GFAP in brain homogenates from human, mouse, rat, bovine, ovine, canine, porcine, rabbit, guinea pig and chicken. 4A11 is particularly useful for identifying GFAP in immunohistochemistry of frozen and formalin-fixed, paraffin-embedded brain tissue sections. Additional applications include western blot analysis and indirect immunofluorescence of tissue-cultured cells. BD Biosciences Pharmingen offers additional GFAP specific antibodies: clone 2E1 (Mfr. No. 556329), 1B4 (Mfr. No. 556328), and clones 1B4, 4A11, 2E1 combined and available as a “cocktail” (Mfr. No. 556330).
Host Species: Mouse
Clone: 4A11
Isotype: IgG2b, κ
Species Reactivity: Human
Immunogen: Cow spinal cord homogenate
Immunofluorescence, Immunohistochemistry, Western Blotting
Specifications
GFAP | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Cow spinal cord homogenate | |
0.5 mg | |
Cell Biology | |
Bovine, Canine, Chicken, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat, Sheep | |
IgG2b |
Immunohistochemistry | |
4A11 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.