missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Bovine Serum Albumin Standard Ampules, 2 mg/mL

High-quality, calibrated BSA solutions (2 mg/mL) to serially dilute to create protein assay standard curves to accurately measure protein concentration.

Brand:  Thermo Scientific™

75.27 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 11811345

  • £97.75 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce BSA Protein Assay Standards are high-quality reference samples for generating accurate standard curves and calibration controls in total protein assays.

These bovine serum albumin (BSA) solutions are protein concentration reference standards for use in BCA, Bradford and other protein assay protocols. BSA is the universally accepted reference protein for total protein quantitation. The albumin standard is precisely formulated at 2mg/mL in an ultrapure 0.9% sodium chloride (saline) solution. The concentration of the stock solution is calibrated by direct comparison to purified BSA (Fraction V) from the National Institute of Standards and Technology (NIST™). The product is available in three convenient package formats: 1mL glass ampules, 50mL polypropylene bottles, and complete sets containing seven ready-to-use dilutions.

Highlights:
Universal – recognized as the industry standard for protein quantitation in colorimetric protein assays
Pure and stable – supplied in ultrapure 0.9% saline solution with 0.05% sodium azide; room temperature stable
Accurate and consistent – precisely formulated at 2.00 +/-0.03mg/mL compared to a NIST reference
Convenient – choose 1mL ampules, a 50mL bottle, or a complete set containing seven ready-to-use dilutions

Includes:
2 mg/mL BSA solution (Cat. No. 23209, 23210) or 7-solution set of different concentrations (Cat. No. 23208)

Recommended for:
Protein assay quantitation standard; Protein recovery control for desalting and other column procedures; General calibration of spectrophotometer UV-lamp (A280)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
Protein Standards
Protein Quantitation Assay
Internal Standard
Pierce Bovine Serum Albumin Standard Ampules, 2mg/mL
Pierce™
Not Target-Specific
10 x 1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ400296 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number23209
Lot NumberZJ400295 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number23209
Lot NumberZI400297 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number23209
Lot NumberZG398161A Certificate TypeCertificate of Analysis Date07/09/2024 Catalog Number23209
Lot NumberZG398161 Certificate TypeCertificate of Analysis Date07/09/2024 Catalog Number23209
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    43

    Hours

    6

    Minutes

    29

    Seconds

    12
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Bovine Serum Albumin Standard Ampules, 2 mg/mL > 2mg/mL solution in glass ampules; 10 x 1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.