missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ MES SDS Running Buffer (20X)

Made with high-purity reagents and are strictly quality controlled

Brand:  Invitrogen™ NP0002

46.51 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 11509166

  • £76.25 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: NuPAGE MES SDS Running Buffer (20X)

Requires : The buffers are provided as a concentrated solution and require simple dilution with deionized water before use.
This item is not returnable. View return policy

Description

Description

NuPAGE MES SDS Running Buffer (20X) is formulated for running proteins on NuPAGE Novex Bis-Tris gels only. NuPAGE MES SDS Running Buffer is recommended for separating small- to medium-sized proteins.

  • Use of MES buffer allows proteins to run faster
  • Ensures high-quality, consistent electrophoresis results

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Running Buffer
7.3
NuPAGE
1X: 50 mM MES, 50 mM Tris Base, 0.1% SDS, 1 mM EDTA, pH 7.3
20X
Liquid
500 mL
Water
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2996843 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberNP0002,NP0002_LITL18 PROMO
Lot Number3070417 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberNP0002,NP0002_LITL18 PROMO
Lot Number3039240 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberNP0002,NP0002_LITL18 PROMO
Lot Number3065314 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberNP0002,NP0002_LITL18 PROMO
Lot NumberTESTCJ23 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog NumberNP0002,NP0002_LITL18 PROMO
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ NuPAGE™ MES SDS Running Buffer (20X) > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.