Learn More
Invitrogen™ SimplyBlue™ SafeStain

SimplyBlue SafeStain is a ready-to-use, fast, sensitive, and safe Coomassie G-250 stain for visualizing protein bands on polyacrylamide gels and on dry PVDF membranes.
Brand: Invitrogen™ LC6060
114.67 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
159.76 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Includes: SimplyBlue™ SafeStain is supplied as a 1X ready-to-use staining reagent.
Description
SimplyBlue SafeStain is a ready-to-use, fast, sensitive, and safe Coomassie G-250 stain for visualizing protein bands on polyacrylamide gels and on dry PVDF membranes. SimplyBlue SafeStain is completely non-hazardous and does not require methanol or acetic acid fixatives or destains. Risk of hazardous exposure and unpleasant odors are eliminated, creating a healthier lab environment.
Coomassie G-250 stain for visualizing protein bands on polyacrylamide gels and on dry PVDF membranes. SimplyBlue SafeStain is completely non-hazardous and does not require methanol or acetic acid fixatives or destains. Risk of hazardous exposure and unpleasant odors are eliminated, creating a healthier lab environment.Features include:
• An easy-to-perform protocol that can be completed in less than three hours. For added speed, a simple microwave procedure can be completed in 12 minutes
• Destaining not required but may be performed with deionized water to achieve maximum sensitivity, especially when performing downstream analyses such as mass spectrometry, or when a crystal-clear background is required
• Sensitive—detect down to 7 ng of protein
• Fast—detect 1 μg of protein in as little as five minutes after staining
• Convenient—save valuable time by skipping reagent preparation with this ready-to-use formulation
• Non-hazardous—save money and hassle associated with hazardous shipping and disposal of hazardous waste
Order Info
Shipping Condition: Room Temperature

Specifications
SimplyBlue™ SafeStain is supplied as a 1X ready-to-use staining reagent. Store at room temperature. Guaranteed stable for 6 months when properly stored. There are no HazMat charges associated with this product. | |
Blue | |
Coomassie | |
Less hazardous | |
1 L |
In-Gel Detection | |
Colorimetric | |
Safe Protein Stain | |
SimplyBlue™ | |
Protein |
Safety and Handling
- SimplyBlue SafeStain
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : citric acid, monohydrate, ethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.