Learn More
Invitrogen™ Colloidal Blue Staining Kit
Five times more sensitivity than Coomassie™ R-250— and can detect as little as 10ng of BSA
Brand: Invitrogen™ LC6025
191.64 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Includes: The Colloidal Blue Staining Kit is supplied with sufficient stainer A and stainer B to stain 25 mini-gels
Description
The Colloidal Blue Staining Kit allows detection of nanogram levels of protein with minimal effort and requires only water to destain. The Colloidal Blue stain uses colloidal chemistry that reduces free dye in solution and improves the protein-to-dye binding ratio. Samples are intensely stained and visible within 3 hours, background staining is virtually eliminated by destaining overnight with water.
- Easy-to-use, mass spectrometry compatible reagents
- Great densitometry results
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
The Colloidal Blue Staining Kit is supplied with sufficient stainer A and stainer B to stain 25 mini-gels. Store the kit at room temperature. The kit is guaranteed for 6 months when properly stored unless otherwise stated in the product documentation. | |
Colorimetric | |
Protein Gel Staining Kit | |
Room Temperature |
In-Gel Detection | |
Coomassie | |
1 kit | |
Protein |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.