Learn More
anti-IgG1, PE, Clone: X40, BD
Brand:  BD Biosciences 340013
Additional Details : Weight : 0.05710kg
 View more versions of this product
Product Code. 15815848
  • £730.00 / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket
Description
Description
The Mouse IgG1 antibody, clone X40, is derived from the hybridization of Sp2/0 mouse myeloma cells with spleen cells isolated from BALB/c mice immunized with KLH. The Mouse IgG1 antibody is specific for keyhole limpet hemocyanin (KLH), an antigen not expressed on human peripheral blood platelets.
Specifications
Specifications
IgG1
X40
50 μg
Immunology
Human
Monoclonal
PE