missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1, PE, Clone: X40, BD
£730.00
Specifications
Antigen | IgG1 |
---|---|
Clone | X40 |
Classification | Monoclonal |
Conjugate | PE |
Regulatory Status | RUO |
Description
Control, Flow Cytometry
Specifications
IgG1 | |
Monoclonal | |
RUO | |
Primary |
X40 | |
PE | |
Immunology |