missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1 κ, Rat, FITC, Clone: R3-34, Isotype Control, BD

Brand:  BD Biosciences 554684

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15847838

  • £1460.00 / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket

Description

Description

The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: R3-34
Isotype: IgG1, κ

Flow Cytometry, Intracellular Staining, Isotype Control

Specifications

Specifications

Flow Cytometry, Western Blot
0.5mg/mL
Rat
0.1 mg
IgG1 κ
R3-34
FITC
Affinity Purified
Undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers