missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1 κ, Rat, FITC, Clone: R3-34, Isotype Control, BD
£275.00 - £1460.00
Specifications
Clone | R3-34 |
---|---|
Applications | Flow Cytometry, Western Blot |
Conjugate | FITC |
Host Species | Rat |
Isotype | IgG1 κ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15847838
|
BD Biosciences
554684 |
0.1 mg |
£1460.00
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
10394114
|
BD Biosciences
553924 |
0.25 mg |
£471.00
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
15866898
|
BD Biosciences
552916 |
50 Tests |
£275.00
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
Description
The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R3-34
Isotype: IgG1, κ
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
R3-34 | |
FITC | |
IgG1 κ | |
Undiluted at 4°C |
Flow Cytometry, Western Blot | |
Rat | |
Affinity Purified |