Learn More
BD anti-Rat Ig, k Light Chain Magnetic Particles - DM IMag-DM, Clone: MRK-1
Brand:  BD 558002
Additional Details : Weight : 0.02000kg
Product Code. 15839158
  • £1355.00 / Each
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
Description
Description
Cell Separation
Specifications
Specifications
Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze.
5 mL
Cell separation