Learn More
Brady™ Anti-skid Laminated Polyester Marking Tape
Anti-Skid (anti-slip) tapes help to prevent slipping and falling on wet or slippery floors. The rugged polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit. Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors. Anti-Skid tape is available in a variety of colors and sizes to suit all of your floor marking needs
Brand:  Brady™ 078147
 View more versions of this product
Product Code. 11761645
  • £99.35 / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The rugged polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit.
  • Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors.
  • Anti-Skid tape is available in a variety of colors and sizes to suit all of your floor marking needs.
Specifications
Specifications
DP-STRIPED ANTI-SKID TAPE BLK/YEL
Striped Anti-Skid Tape - Black / Yellow
Traction on steps, slick floors, and egress pathway marking
18.29 m
-40°F to +158°F
0.162 mm
50.8 mm
Black and Yellow Striped
Anti-skid Tape
ANSI A1264.2-2006, NSFI Certified for High Traction, OSHA 1910.144, OSHA 1910.22
18.29 m x 50.80 mm
60 ft.
1 Roll
-40°C to +70°C
2 in.
Permanent Acrylic
Polyester