missing translation for 'onlineSavingsMsg'
Learn More

Day Impex™ LabGUARD Antimicrobial Hand Soap

Gently but effectively destroy a broad spectrum of organisms. Day Impex™ LabGUARD Antimicrobial Hand Soap is a concentrated coconut based lotion, specially formulated for frequent or repeated use without drying the skin.

£355.00 - £442.00

Specifications

Product Type Hand Soap
Composition Coconut Based Lotion
For Use With (Application) Use as Shower Soap, Combating the Effects of Water, Harsh Solutions and Other Irritants Found in the Laboratory
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
12398657
View Documents
Day Impex™
331002
1 L
£355.00
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
12308667
View Documents
Day Impex™
331003
5 L
£442.00
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Concentrated
  • Coconut-based lotion
  • Safe to use as a shower soap
  • Natural moisturisers and conditioners 
  • Combats the effects of water, harsh solutions and other irritants found in the laboratory
Specifications

Specifications

Hand Soap
Coconut Based Lotion
Use as Shower Soap, Combating the Effects of Water, Harsh Solutions and Other Irritants Found in the Laboratory
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Day Impex™ LabGUARD Antimicrobial Hand Soap

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.