missing translation for 'onlineSavingsMsg'
Learn More
Antimony(III) chloride, ACS, 99.0% min, Thermo Scientific Chemicals Product Code.: 11393348

Antimony(III) chloride, ACS, 99.0% min, Thermo Scientific Chemicals

Product Code. 11393348
250 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11393348

Brand: Thermo Scientific Alfa Aesar 036281.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols and carotenoids. It is likewise employed as a dye mordant and a fabric flame retardant. It is used as inorganic and organic chlorination catalyst. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.5°C
Melting Point 73.4°C
Color White
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 99% min.
Solubility Information Soluble in water,HCl solution,alcohol,benzene,carbon disulfide,dioxane,chloroform,ether,acetone,carbon tetrachloride. Insoluble in organic bases.
Formula Weight 228.11
Grade ACS
Odor Pungent
Sensitivity Moisture sensitive
Appearance White crystalline
Density 3.14
Chemical Name or Material Antimony(III) chloride, For analysis ACS
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM11K008 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number036281.18,036281.A1,036281.30
Lot NumberM08K004 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number036281.18,036281.A1,036281.30
Lot NumberQ12H048 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number036281.18,036281.A1,036281.30
Lot NumberG19M04 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036281.18,036281.A1,036281.30
Lot NumberL01Q084 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036281.18,036281.A1,036281.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony(III) chloride, ACS, 99.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.