Learn More
Antimony trichloride, ACS reagent, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 223.0°C |
Melting Point | 73.0°C |
Color | White |
Physical Form | Powder, Crystals and/or Chunks |
Quantity | 500 g |
Linear Formula | SbCl3 |
Fieser | 08,19 |
Merck Index | 15,696 |
Solubility Information | Solubility in water: 100g/L (25°C). Other solubilities: soluble in diluted hcl,, soluble in chloroform (+/- 22%), soluble in alcohol, benzene, carbon disulfide,, acetone, tartaric acid, ccl4, insoluble in pyridine, quinoline, organic bases |
IUPAC Name | antimony(3+) trichloride |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467864 | Certificate of Analysis | 31/01/2025 | 401370050,401375000,401370025,401371000 |
A0463455 | Certificate of Analysis | 03/07/2024 | 401370050,401375000,401370025,401371000 |
A0416565 | Certificate of Analysis | 12/02/2024 | 401370050,401375000,401370025,401371000 |
A0444924 | Certificate of Analysis | 25/08/2023 | 401370050,401375000,401370025,401371000 |
A0403689 | Certificate of Analysis | 13/12/2022 | 401370050,401375000,401370025,401371000 |
Safety and Handling



- Antimony trichloride
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Antimony trichloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.