Learn More
Antimony(III) isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals
£93.90 - £342.00
Chemical Identifiers
CAS | 18770-47-3 |
---|---|
Molecular Formula | C9H21O3Sb |
Molecular Weight (g/mol) | 299.024 |
MDL Number | MFCD00143623 |
InChI Key | HYPTXUAFIRUIRD-UHFFFAOYSA-N |
Synonym | antimony iii isopropoxide, antimony 3+ tripropan-2-olate, triisopropoxyantimony, acmc-209epx, antimony iii propan-2-olate, antimony 3+ ; propan-2-olate, 2-propanol,antimony 3+ salt 9ci, antimony 3+ ion tris propan-2-olate, antimony 3+ tris propan-2-olate |
PubChem CID | 3034303 |
IUPAC Name | antimony(3+);propan-2-olate |
SMILES | CC(C)[O-].CC(C)[O-].CC(C)[O-].[Sb+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358028
|
Thermo Scientific Alfa Aesar
036589.06 |
5 g |
£93.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348028
|
Thermo Scientific Alfa Aesar
036589.14 |
25 g |
£342.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18770-47-3 | |
299.024 | |
HYPTXUAFIRUIRD-UHFFFAOYSA-N | |
3034303 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Sb+3] |
C9H21O3Sb | |
MFCD00143623 | |
antimony iii isopropoxide, antimony 3+ tripropan-2-olate, triisopropoxyantimony, acmc-209epx, antimony iii propan-2-olate, antimony 3+ ; propan-2-olate, 2-propanol,antimony 3+ salt 9ci, antimony 3+ ion tris propan-2-olate, antimony 3+ tris propan-2-olate | |
antimony(3+);propan-2-olate |
Specifications
18770-47-3 | |
1.31 g/mL | |
77°C (141°F) | |
MFCD00143623 | |
Moisture sensitive | |
HYPTXUAFIRUIRD-UHFFFAOYSA-N | |
antimony(3+);propan-2-olate | |
3034303 | |
99.9% | |
(metals basis) |
Colorless | |
68°C (3 mmHg) | |
C9H21O3Sb | |
UN3082 | |
antimony iii isopropoxide, antimony 3+ tripropan-2-olate, triisopropoxyantimony, acmc-209epx, antimony iii propan-2-olate, antimony 3+ ; propan-2-olate, 2-propanol,antimony 3+ salt 9ci, antimony 3+ ion tris propan-2-olate, antimony 3+ tris propan-2-olate | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Sb+3] | |
299.024 | |
298.8 | |
Liquid | |
Antimony(III) isopropoxide |
Safety and Handling
GHS H Statement
H302-H332-H227
Harmful if swallowed.
Harmful if inhaled.
Combustible liquid.
P210-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
H227-H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 242-561-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only