Learn More
Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
£26.60 - £282.00
Chemical Identifiers
CAS | 28300-74-5 |
---|---|
Molecular Formula | C8H4K2O12Sb2·3H2O |
MDL Number | MFCD00148863 |
Synonym | Potassium antimonyl tartrate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11410321
|
Thermo Scientific Alfa Aesar
040619.14 |
25 g |
£26.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11378568
|
Thermo Scientific Alfa Aesar
040619.22 |
100 g |
£72.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388568
|
Thermo Scientific Alfa Aesar
040619.36 |
500 g |
£282.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Antimony potassium tartrate trihydrate is used as a antiparasitic agent, which used in the treatment of schistosomiasis and leishmaniasis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
28300-74-5 | |
C8H4K2O12Sb2·3H2O | |
MFCD00148863 | |
Potassium antimonyl tartrate |
Specifications
28300-74-5 | |
99.0-103.0% | |
MFCD00148863 | |
Soluble in water and glycerine. Insoluble in ethanol. | |
ACS | |
Antimony potassium tartrate trihydrate |
Powder | |
C8H4K2O12Sb2·3H2O | |
Potassium antimonyl tartrate | |
667.87 (613.82 Anhydrous) | |
2.6 |
Safety and Handling
Avoid release to the environment. Refer to special instructions/Safety data sheets
P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
H302+H332-H335-H373
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 229-436-1
missing translation for 'rtecsNumber' : CC6825000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only