Learn More
Antimony shot, 1.5-5mm (0.06-0.2in), Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
£34.37 - £354.77
Chemical Identifiers
CAS | 7440-36-0 |
---|---|
Molecular Formula | Sb |
Molecular Weight (g/mol) | 121.76 |
MDL Number | MFCD00011211 MFCD00134030 |
InChI Key | WATWJIUSRGPENY-UHFFFAOYSA-N |
Synonym | powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes germanShow More |
PubChem CID | 5354495 |
ChEBI | CHEBI:30304 |
SMILES | [Sb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346567
|
Thermo Scientific Alfa Aesar
000428.06 |
5 g |
£34.37
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336567
|
Thermo Scientific Alfa Aesar
000428.14 |
25 g |
£107.54
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326567
|
Thermo Scientific Alfa Aesar
000428.22 |
100 g |
£354.77
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
7440-36-0 | |
121.76 | |
WATWJIUSRGPENY-UHFFFAOYSA-N | |
5354495 | |
[Sb] |
Sb | |
MFCD00011211 MFCD00134030 | |
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes germanShow More | |
CHEBI:30304 |
Spezifikation
Silver to Gray | |
Solid in various forms | |
Sb | |
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german | |
[Sb] | |
121.76 | |
CHEBI:30304 | |
Shot |
7440-36-0 | |
99.9999% (Metals basis) | |
MFCD00011211 MFCD00134030 | |
WATWJIUSRGPENY-UHFFFAOYSA-N | |
distibyne | |
5354495 | |
Odorless | |
Antimony |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 231-146-5
missing translation for 'rtecsNumber' : CC4025000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.