Learn More
Antimony(V)-chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 434528000
264.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Antimony(V)-chloride | |
Yellow | |
800 mL | |
SbCl5 | |
15,686 | |
VMPVEPPRYRXYNP-UHFFFAOYSA-I | |
pentachloro-$l^{5}-stibane | |
24294 | |
AcroSeal™ Glass Bottle | |
1.4420g/mL |
7647-18-9, 75-09-2 | |
Solution | |
Cl5Sb | |
MFCD00011213 | |
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german | |
Cl[Sb](Cl)(Cl)(Cl)Cl | |
299.02 | |
299.01 | |
1.442 |
Safety and Handling
- Antimony(V) chloride, 1M solution in methylene chloride
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only