Learn More
Antimony(V)-chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
£61.04 - £273.36
Specifications
Chemical Name or Material | Antimony(V)-chloride |
---|---|
CAS | 7647-18-9, 75-09-2 |
Color | Yellow |
Physical Form | Solution |
Molecular Formula | Cl5Sb |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
15721819
|
Thermo Scientific Acros
434521000 |
100 mL | AcroSeal™ Glass Bottle |
£61.04
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11924201
|
Thermo Scientific Acros
434528000 |
800 mL | AcroSeal™ Glass Bottle | N/A | N/A | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Antimony(V)-chloride | |
Yellow | |
Cl5Sb | |
MFCD00011213 | |
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german | |
Cl[Sb](Cl)(Cl)(Cl)Cl | |
299.02 | |
299.01 | |
1.442 |
7647-18-9, 75-09-2 | |
Solution | |
SbCl5 | |
15,686 | |
VMPVEPPRYRXYNP-UHFFFAOYSA-I | |
pentachloro-$l^{5}-stibane | |
24294 | |
AcroSeal™ Glass Bottle | |
1.4420g/mL |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-601-8
- Antimony(V) chloride, 1M solution in methylene chloride
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Dichloromethane, Antimony pentachloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.