Learn More
Antimycin A, Thermo Scientific Chemicals
£57.10 - £81.10
Chemical Identifiers
CAS | 1397-94-0 |
---|---|
Molecular Formula | C28H40N2O9 |
Molecular Weight (g/mol) | 548.633 |
MDL Number | MFCD01779723 |
InChI Key | UIFFUZWRFRDZJC-RBVQMQRASA-N |
PubChem CID | 16218979 |
IUPAC Name | [(2R,3S,6S,7R)-3-[(3-formamido-2-hydroxybenzoyl)amino]-8-hexyl-2,6-dimethyl-4,9-dioxo-1,5-dioxonan-7-yl] 3-methylbutanoate |
SMILES | CCCCCCC1C(C(OC(=O)C(C(OC1=O)C)NC(=O)C2=C(C(=CC=C2)NC=O)O)C)OC(=O)CC(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15495729
|
Thermo Scientific Alfa Aesar
J63522.#0 |
5 mg |
£57.10
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15405739
|
Thermo Scientific Alfa Aesar
J63522.MA |
10 mg |
£81.10
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAntimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.
Solubility
Soluble in 100% ethanol. Insoluble in water.
Notes
Isolated from Streptomyces sp.
Chemical Identifiers
1397-94-0 | |
548.633 | |
UIFFUZWRFRDZJC-RBVQMQRASA-N | |
[(2R,3S,6S,7R)-3-[(3-formamido-2-hydroxybenzoyl)amino]-8-hexyl-2,6-dimethyl-4,9-dioxo-1,5-dioxonan-7-yl] 3-methylbutanoate |
C28H40N2O9 | |
MFCD01779723 | |
16218979 | |
CCCCCCC1C(C(OC(=O)C(C(OC1=O)C)NC(=O)C2=C(C(=CC=C2)NC=O)O)C)OC(=O)CC(C)C |
Specifications
Antimycin A | |
1397-94-0 | |
C28H40N2O9 | |
UN3462 | |
Soluble in 100% ethanol. Insoluble in water. | |
CCCCCCC1C(C(OC(=O)C(C(OC1=O)C)NC(=O)C2=C(C(=CC=C2)NC=O)O)C)OC(=O)CC(C)C | |
548.633 | |
548.63 | |
Light sensitive |
Powder | |
149°C to 150°C | |
MFCD01779723 | |
14,714 | |
UIFFUZWRFRDZJC-RBVQMQRASA-N | |
[(2R,3S,6S,7R)-3-[(3-formamido-2-hydroxybenzoyl)amino]-8-hexyl-2,6-dimethyl-4,9-dioxo-1,5-dioxonan-7-yl] 3-methylbutanoate | |
16218979 | |
White to Yellow | |
Antimycin A |
Safety and Handling
GHS H Statement
H300
Fatal if swallowed.
P262-P264b-P270-P280-P301+P310-P302+P350-P310-P330-P361-P363-P501c
H300+H310
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.
missing translation for 'rtecsNumber' : CD0350000
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only