missing translation for 'onlineSavingsMsg'
Learn More
Antimycin A, Thermo Scientific Chemicals Product Code.: 15405739

Antimycin A, Thermo Scientific Chemicals

Product Code. 15405739
10 mg
Click to view available options
Quantity:
5 mg
10 mg
This item is not returnable. View return policy

Product Code. 15405739

Brand: Thermo Scientific Alfa Aesar J63522.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.

Solubility
Soluble in 100% ethanol. Insoluble in water.

Notes
Isolated from Streptomyces sp.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1397-94-0
Molecular Formula C28H40N2O9
Molecular Weight (g/mol) 548.633
MDL Number MFCD01779723
InChI Key UIFFUZWRFRDZJC-RBVQMQRASA-N
PubChem CID 16218979
IUPAC Name [(2R,3S,6S,7R)-3-[(3-formamido-2-hydroxybenzoyl)amino]-8-hexyl-2,6-dimethyl-4,9-dioxo-1,5-dioxonan-7-yl] 3-methylbutanoate
SMILES CCCCCCC1C(C(OC(=O)C(C(OC1=O)C)NC(=O)C2=C(C(=CC=C2)NC=O)O)C)OC(=O)CC(C)C

Specifications

Type Antimycin A
Physical Form Powder
Melting Point 149°C to 150°C
Color White to Yellow
Quantity 10 mg
Sensitivity Light sensitive
UN Number UN3462
Merck Index 14,714
Solubility Information Soluble in 100% ethanol. Insoluble in water.
Formula Weight 548.63
Chemical Name or Material Antimycin A
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP05L008 Certificate TypeCertificate of Analysis Date07/03/2025 Catalog NumberJ63522.#0,J63522.MA
Lot NumberN28L021 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberJ63522.#0,J63522.MA
Lot NumberZ20K018 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog NumberJ63522.#0,J63522.MA
Lot NumberY06K025 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberJ63522.#0,J63522.MA
Lot NumberX15K050 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberJ63522.#0,J63522.MA
compliance-icons compliance-icons
Product Identifier
  • Antimycin A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : Antimycin A

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimycin A, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.