missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin Sulfate (White Powder), Fisher BioReagents

For selecting transformed cells containing kanamycin resistance gene

Brand:  Fisher BioReagents BP906-5

65.64 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.



Product Code. 10031553

  • £90.05 / 5g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

For selecting transformed cells containing kanamycin resistance gene

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kanamycin Sulfate
Solid
6.5 to 8.5
Poly Bottle
White
C18H38N4O15S
OOYGSFOGFJDDHP-IZQIRFRQSA-N
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
134129479
Kanamycin Sulfate
25389-94-0
< 1.0%
> 750μg/mg
5 g
15, 5329
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
582.575
582.58
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kanamycin Sulfate (White Powder), Fisher BioReagents > Poly Bottle; 5g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.