Learn More
MP Biomedicals™ Vitamin Diet Fortification Mixture
VDFM
Brand: MP Biomedicals™ 0290465401
Description
MP Vitamin Diet Fortification Mixture formulation provides sufficient vitamin levels to prevent deficiencies in all animal species, giving precise control of vitamin levels.

Specifications
Animal Diet | |
Powder | |
1 kg | |
15°C to 30°C |
Vitamin A Acetate (500000 IU/g) 1.8 g/k g, Vitamin D2 (850000 IU/g) 0.125 g/k g, DL-a-Tocopherol Acetate 22 g/k g, Ascorbic Acid 45 g/k g, Inositol 5 g/k g, Choline Chloride 75 g/k g, Menadione 2.25 g/k g, p-Aminobenzoic Acid 5 g/k g, Niacin 4.25 g/k g, Riboflavin 1 g/k g, Pyridoxine Hydrochloride 1 g/k g, Thiamine Hydrochloride 1 g/k g, Calcium Pantothenate 3 g/k g, Biotin 0.02 g/k g, Folic Acid 0.09 g/k g, Vitamin B12 0.00135 g/k g | |
MP Vitamin Diet fortification mixture formulation provides sufficient Vitamin levels to prevent deficiencies in all animal species, giving precise control of Vitamin levels | |
Rat |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- MP Vitamin Diet Fortification Mixture
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1A
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.