Learn More
Grant Instruments™ ap360 Air Purifier
Grant Instruments' core engineering strengths and manufacturing expertise, together with proven science around ultraviolet light (UVC) as a disinfectant, mean our quiet, high-performance ap360 Air Purifier device is an ideal air hygiene solution for clinical settings and businesses
Brand: Grant Instruments™ AP360
This item is not returnable.
View return policy
Description
- Grant Instruments ap360 Air Purifier is designed to help you keep your staff, patients, and customers safe from airborne infection as well as to remove aerosols, pollen, dust and other particulates
- UVC lamps provide powerful UV air sterilisation that disrupts the DNA and RNA in bacteria and viruses
- Large air processing capacity of 360m3/hour enables minimal downtime
- Fast fan speed with three modes and auto settings
- Digital timer 1 min to 24 hrs/non-stop can be set to the desired minimum or maximum use
- Full user protection from direct UV light
- Compact design to minimise use of space and maximise portability
- Low power usage for a cost-effective and eco-friendly air purification solution
Specifications
0.9995 | |
360 m3/h | |
1050 mm | |
350 mm | |
120/230 V | |
EN1822 Standard HEPA13 |
HEPA13 | |
13.4 Kg | |
350 mm | |
Air Purifier | |
50/60 Hz |
Product Content Correction
Product Title
Grant Instruments™ ap360 Air Purifier > Grant Instruments ap360 Air Purifier
Spot an opportunity for improvement?Share a Content Correction