Learn More
A&D Weighing Apollo Precision Balance Class II Approved
GREENER_CHOICE
Apollo Analytical Series improves productivity and efficiency of your workflows with user-friendly and innovative features. It is designed to provide reliable results across a range of environments and applications.
£1600.00 - £2300.00
Specifications
Certifications/Compliance GLP/GMP/GCP/ISO/OIML
Connection Type RS-232 C and USB
Display Type Reverse-backlit LCD
Material Stainless Steel
Stabilization Time 1 sec.
View More Specs
Products 9
Product Code Brand Capacity Readability Price Quantity & Availability  
Product Code Brand Capacity Readability Price Quantity & Availability  
16864601
GREENER_CHOICE
View Documents
A&D Weighing
GX-203A-EC
220 g 0.001 g
£1805.00
Each
16804611
GREENER_CHOICE
View Documents
A&D Weighing
GX-303A-EC
320 g 0.001 g
£1905.00
Each
16844611
GREENER_CHOICE
View Documents
A&D Weighing
GX-403A-EC
420 g 0.001 g
£1975.00
Each
16874611
GREENER_CHOICE
View Documents
A&D Weighing
GX-603A-EC
620 g 0.001 g
£2300.00
Each
16854601
GREENER_CHOICE
View Documents
A&D Weighing
GX-2002A-EC
2200 g 0.01 g
£1785.00
Each
16894601
GREENER_CHOICE
View Documents
A&D Weighing
GX-3002A-EC
3200 g 0.01 g
£1915.00
Each
16834611
GREENER_CHOICE
View Documents
A&D Weighing
GX-4002A-EC
4200 g 0.01 g
£1975.00
Each
16864611
GREENER_CHOICE
View Documents
A&D Weighing
GX-6002A-EC
6200 g 0.01 g
£2300.00
Each
16854611
GREENER_CHOICE
View Documents
A&D Weighing
GX-6001A-EC
6200 g 0.1 g
£1600.00
Each
Beschreibung
Beschreibung
  • Newly developed Smart SHS weighing sensor featuring ECL technology
  • APA (Auto Precision Assessment) – Automatic assessment of the balance performance, calculation and display of Standard Deviation (repeatability) at the point of use without the need for calibration weights, can be checked daily via a key press
  • Automatic Minimum Weight Calculation and setting via APA function
  • Input minimum weight automatically via APA or manually using weights
  • Visual Minimum Weight alert
  • UAC (User Access Control) – Administrator and Operator password access to ensure data integrity
  • Calibration via One-touch, Interval, pre-set times (3) or automatically in response to ambient temperature changes (GX-AE/A)
  • ISD (Patented Impact Shock Detection) – 4 level shock detection with visual and audible feedback, maintain the performance of your investment through correct use of the balance, ideal in automated loading applications and ideal for operator guidance
  • FRD (Flow Rate Display) – Direct display of flow rate of a sample in g/s, g/m, g/h or volume flow rate in mL/s, mL/m, mL/h
  • Up to 10 sample densities can be saved to memory
  • Use in conjunction with the flow Rate Comparator to keep flow rates within designated limits
  • SCF (Statistical Calculation Function) – Display and print various statistical results on the balance display or output to a printer or PC
  • Standard RS232 and Dual Mode USB interface. USB can be configured for quick mode (connect to PC without the need for software installation) or bi-directional mode
  • UFC (Universal Flexi Coms) – configurable data output, ideal for connection to data acquisition systems or barcode label printers etc.
  • Gross/Net/Tare data output, Tare memory, Underhook, Capacity Indicator
  • Large selection of Options and Accessories
  • Built-in fanless ionizer with Quick Ion™ technology and external IR switch (GX-AE)
Spezifikation
Spezifikation
GLP/GMP/GCP/ISO/OIML
Reverse-backlit LCD
1 sec.
RS-232 C and USB
Stainless Steel
30 VA to adapter (12 V, 1 A)
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
A&D Weighing Apollo Precision Balance Class II Approved
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt