Learn More
Applied Biosystems™ AmpliTaq™ 360 DNA Polymerase
The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts.
Brand: Applied Biosystems™ 4398891
Code : 50
Additional Details : Weight : 0.13000kg
Description
The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts. Compared to the original AmpliTaq DNA polymerase, AmpliTaq 360 polymerase is purified by an additional proprietary separation process which reduces contaminating bacterial DNA sequences from the enzyme preparation. This ultra-pure enzyme reduces false positive results, amplifies low-level target sequences, and when combined with the proprietary AmpliTaq 360 Buffer Kit, promotes the amplification of a variety of templates including those from bacterial and human genomes.
- Optimized for the broadest range of targets—from everyday to challenging
- Unmatched sensitivity and yield
- Market-leading GC enhancer for robust amplification of GC-rich sequences
- Achieves the highest quality sequencing data
For superior PCR performance, we recommend DreamTaq DNA Polymerase.
Specifications
• 10X AmpliTaq 360 PCR Buffer, 1.5 mL for each 250 U of enzyme All solutions are prepared in deionized, filter-sterilized water. |
|
Medium | |
5' - 3' | |
5 U/μL | |
3'-A | |
5 kb or less |
Stand-alone enzyme | |
AmpliTaq 360 DNA Polymerase | |
1500 Units | |
1X | |
Separate Components | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.