missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYBR™ Power SYBR™ Green PCR Master Mix
Offers superior sensitivity and reproducibility, detecting as few as two copies of a target gene over a broad range of template concentrations.
Brand: SYBR™ 4368708
1559.25 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Includes: SYBR™ Green 1 Dye, AmpliTaq Gold DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components
Requires : Template and primers
This item is not returnable. View return policy
Description
Get superior nucleic acid quantitation with Applied Biosystems™ Power SYBR™ Green Master Mix.
- Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient single-tube format
- Power SYBR™ Green PCR Master Mix contains all of the components (excluding the template and primers) for superior SYBR™ Green reagent-based real-time PCR
- The optimized formulation contains highly purified AmpliTaq Gold™ DNA Polymerase, LD to offer increased sensitivity
- Contains a blend of dTTP/dUTP that is compatible with AmpErase™ UNG to minimize carryover contamination
- Includes a proprietary version of ROX™ dye, an internal passive reference, to normalize non-PCR–related fluorescence fluctuations
- This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error and sample evaporation
- Power SYBR™ Green reagent-based PCR chemistry easily replaces the existing SYBR™ Green PCR Master Mix in Applied Biosystems™ protocols using the same setup and thermal cycling conditions
- Store at -20°C
Order Info
Shipping Condition: Wet Ice
Specifications
The 2X mix contains SYBR™ Green 1 Dye, AmpliTaq Gold™ DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components. Contains 10 × 5 mL bottles, sufficient for 2000 reactions of 50 μL each. Store at -20°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Tube | |
qPCR | |
Standard | |
SYBR Green | |
SYBR™ | |
LD (Low DNA) | |
2000 Reactions of 50 μL each | |
2X | |
DNA (Genomic), cDNA |
SYBR | |
High | |
AmpliTaq Gold DNA Polymerase, AmpliTaq Gold LD | |
7300 System, 7500 System, 7700 System, 7900HT Fast System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
10 x 5 mL | |
Gene Expression | |
2000 Reactions | |
10 x 5 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction