missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 48-Well Optical Adhesive Film

Reduces the chance of well-to-well contamination and sample evaporation when applied to a 48-well microplate

Brand:  Applied Biosystems™ 4375323

168.79 GBP valid until 2024-12-31
Use promo code "21898" to get your promotional price.



Code : 50

Additional Details : Weight : 0.14000kg

 View more versions of this product

Product Code. 10488444

  • £201.00 / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

  • Pressure-sensitive adhesive provides tight seal over every well
  • Will not interfere with sample reading
  • Easy to apply and remove, will not stick to gloves
  • Prevent Sample Loss
  • Optically clear adhesive film prevents sample loss by securely sealing into wells of microplates and reducing leaks, evaporation, and contamination between sample wells to help ensure consistent real-time PCR data
  • Easy to Use: simply peel backing and apply firmly across top of microplate
  • Once sealed, microplates can be safely stacked without worrying about contamination
  • Designed to fit MicroAmp Fast Optical 48-Well Plates
Note: See user's manual or package insert for limited label license and trademark information.

For Research Use Only. Not for use in diagnostic procedures.

Applications:

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Specifications

Specifications

Optical
48
StepOne™,
Film, Seal, or Applicator
Pressure Sensitive
Polyester
Contains 100 MicroAmp™ 48-Well Optical Adhesive Films. Store at room temperature.
Room Temperature
MicroAmp
48-well Plate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers