missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 48-Well Optical Adhesive Film

Reduces the chance of well-to-well contamination and sample evaporation when applied to a 48-well microplate

Brand:  Applied Biosystems™ 4375928

31.88 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10784027

  • £62.50 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: 100 or 25 MicroAmp 48-Well Optical Adhesive Films
This item is not returnable. View return policy

Description

Description

  • Pressure-sensitive adhesive provides tight seal over every well
  • Will not interfere with sample reading
  • Easy to apply and remove, will not stick to gloves
  • Prevent Sample Loss
  • Optically clear adhesive film prevents sample loss by securely sealing into wells of microplates and reducing leaks, evaporation, and contamination between sample wells to help ensure consistent real-time PCR data
  • Easy to Use: simply peel backing and apply firmly across top of microplate
  • Once sealed, microplates can be safely stacked without worrying about contamination
  • Designed to fit MicroAmp Fast Optical 48-Well Plates
Note: See user's manual or package insert for limited label license and trademark information.

For Research Use Only. Not for use in diagnostic procedures.

Applications:

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Optical
48
Contains 25 MicroAmp™ 48-Well Optical Adhesive Films. Store at room temperature.
Room Temperature
MicroAmp
48-well Plate
Polyester
25 films
StepOne™,
Film, Seal, or Applicator
Pressure Sensitive
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.