Learn More
SYBR™ Fast SYBR™ Green Master Mix
Designed for speed, SYBR™ Green Master Mix provides a fast, reliable, and cost-effective solution for RT-PCR application without compromising sensitivity, specificity, dynamic range, or PCR efficiency.
Brand: Applied Biosystems™ 4385616
667.65 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
- A fast, reliable, and cost-effective solution for real-time PCR applications without compromising sensitivity, specificity, dynamic range, or PCR efficiency
- Real-time PCR results in as fast as 35 minutes
- Capable of detecting very low copies of target
- Minimizes primer-dimer and non-specific amplification
- Consistent amplification across a wide dynamic range
- Contains all of the components, excluding the template and primers, in a convenient 2X master mix
- Store at -5°C to -30°C
Includes the following components in an optimized buffer:
- AmpliTaq™ Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start
- Minimizes non-specific product formation and enables reactions to be set up at room temperature
- SYBR™ Green I dye to enable detection of double-stranded DNA
- Deoxynucleotides (dNTPs) to help maintain optimal PCR results
- Uracil-DNA Glycosylase (UDG) designed to reduce carryover contamination
- Passive internal reference based on proprietary ROX™ dye to enable increased precision
Specifications
Contains 2 × 5 mL vials. The 2X mix contains SYBR™ Green 1 Dye, AmpliTaq Gold™ Fast DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components. Sufficient for 1000 reactions of 20 μL each. Store at -5 to -30°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Vial | |
qPCR | |
Fast | |
SYBR Green | |
SYBR™ | |
UP (Ultra Pure) | |
1000 Reactions of 20 μL Each | |
2X | |
DNA (Genomic), cDNA |
SYBR | |
High | |
AmpliTaq Fast DNA Polymerase, AmpliTaq Fast UP | |
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
2 x 5 mL | |
Gene Expression | |
1000 Reactions | |
2 x 5 mL |
For Research Use Only. Not for use in diagnostic procedures.