missing translation for 'onlineSavingsMsg'
Learn More

SYBR™ SYBR™ Select Master Mix for CFX

A cost-effective solution for real-time PCR applications.

Brand:  SYBR™ 4472954

2618.35 GBP valid until 2024-12-31
Use promo code "21881" to get your promotional price.


 View more versions of this product

Product Code. 13246529

  • £3040.00 / 5000 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 2X mix contains SYBR GreenER dye, AmpliTaq DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, and optimized buffer components.
This item is not returnable. View return policy

Description

Description

  • Specific - minimize primer-dimer and non-specific amplification
  • Reproducible and sensitive - consistent amplification across a wide dynamic range
  • Bright - contains SYBR™ GreenER™ dye for maximum brightness
  • Carry-over contamination control - contains heat-labile UDG
  • For use with the Bio-Rad CFX96™ Touch or CFX384™ Touch systems
  • Standard and fast cycling parameters

Formulated for Maximum Specificity:
  • SYBR™ Select Master Mix for CFX contains all the components needed for your real-time PCR reactions, except the template and primers, in a convenient 2X concentration premix
  • The master mix includes AmpliTaq™ DNA Polymerase, UP, a highly purified DNA polymerase with a proprietary hot start mechanism that provides exceptional specificity

Obtain Reproducible Results Across a Wide Dynamic Range:
  • SYBR™ Select Master Mix for CFX is specially formulated to provide robust results from 100ng to 0.1pg cDNA per reaction and can reliably detect a single copy of genomic DNA.
    • Contains SYBR™ GreenER™ Dye and Heat-labile UDG:
    • SYBR™ GreenER™ dye is less inhibitory to PCR than SYBR™ Green I dye, resulting in brighter signals
    • Heat-labile uracil-DNA glycosylase (UDG) is included for worry-free carryover contamination control

    Order Info

    Shipping Conditions: Wet Ice

    TRUSTED_SUSTAINABILITY
Specifications

Specifications

The 2x mix contains SYBR™ GreenER™ dye, AmpliTaq™ DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, and optimized buffer components. Contains 10 X 5 mL bottles, sufficient for 5000 reactions at 20 μL. Store at 2-8™C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Liquid
High
AmpliTaq DNA Polymerase
AB 7900HT, Standard Mode, AB StepOne™, Fast Mode, AB 7500, Standard Mode, AB 7500, Fast Mode, Bio-Rad CFX96 Touch™, AB StepOnePlus™, Fast Mode, AB 7900HT, Fast Mode, AB StepOne™, Standard Mode, Bio-Rad CFX Connect™, AB 7900 HT 384-Well System, Bio-Rad CFX384 Touch™, AB StepOnePlus™, Standard Mode
ROX (Pre-mixed)
Real Time PCR SYBR Master Mix
10 x 5 mL
Gene Expression
Built-In Hot Start
DNA (Genomic), cDNA
SYBR
Vial
qPCR
Fast or Standard
SYBR GreenER
SYBR™
UP (Ultra Pure)
5000 Reactions of 20 μL
2X
5000 Reactions
10 x 5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.