missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYBR™ SYBR™ Select Master Mix for CFX
A cost-effective solution for real-time PCR applications.
£87.25 - £3040.00
Specifications
Detection Method | SYBR |
---|---|
Form | Liquid |
Format | Vial |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Includes: 2X mix contains SYBR GreenER dye, AmpliTaq DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, and optimized buffer components.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13296519
|
SYBR™
4472937 |
1 x 1 mL |
£87.25
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13206529
|
SYBR™
4472942 |
1 x 5 mL |
£365.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13216529
|
SYBR™
4472952 |
2 x 5 mL |
£680.00
1000 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13246529
|
SYBR™
4472954 |
10 x 5 mL |
£3040.00
5000 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13226529
|
SYBR™
4472953 |
5 x 5 mL |
£1570.00
2500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Specific - minimize primer-dimer and non-specific amplification
- Reproducible and sensitive - consistent amplification across a wide dynamic range
- Bright - contains SYBR™ GreenER™ dye for maximum brightness
- Carry-over contamination control - contains heat-labile UDG
- For use with the Bio-Rad CFX96™ Touch or CFX384™ Touch systems
- Standard and fast cycling parameters
Formulated for Maximum Specificity:
- SYBR™ Select Master Mix for CFX contains all the components needed for your real-time PCR reactions, except the template and primers, in a convenient 2X concentration premix
- The master mix includes AmpliTaq™ DNA Polymerase, UP, a highly purified DNA polymerase with a proprietary hot start mechanism that provides exceptional specificity
Obtain Reproducible Results Across a Wide Dynamic Range:
- SYBR™ Select Master Mix for CFX is specially formulated to provide robust results from 100ng to 0.1pg cDNA per reaction and can reliably detect a single copy of genomic DNA.
- Contains SYBR™ GreenER™ Dye and Heat-labile UDG:
- SYBR™ GreenER™ dye is less inhibitory to PCR than SYBR™ Green I dye, resulting in brighter signals
- Heat-labile uracil-DNA glycosylase (UDG) is included for worry-free carryover contamination control
Order Info
Shipping Conditions: Wet Ice
Specifications
SYBR | |
Vial | |
qPCR | |
Fast or Standard | |
SYBR GreenER | |
SYBR™ | |
UP (Ultra Pure) | |
2X | |
DNA (Genomic), cDNA |
Liquid | |
High | |
AmpliTaq DNA Polymerase | |
AB 7900HT, Standard Mode, AB StepOne™, Fast Mode, AB 7500, Standard Mode, AB 7500, Fast Mode, Bio-Rad CFX96 Touch™, AB StepOnePlus™, Fast Mode, AB 7900HT, Fast Mode, AB StepOne™, Standard Mode, Bio-Rad CFX Connect™, AB 7900 HT 384-Well System, Bio-Rad CFX384 Touch™, AB StepOnePlus™, Standard Mode | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
Built-In Hot Start |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title