Learn More
Applied Biosystems™ TaqMan™ Reverse Transcription Reagents
TaqMan Reverse Transcription Reagents provide the necessary reagents for reverse transcriptase-PCR (RT-PCR). Random hexamers, oligo (dT)16, and reverse primers are included for cDNA synthesis flexibility.
£506.00 - £4290.00
Specifications
Format | Kit |
---|---|
Technique | Reverse Transcription |
Optimal Reaction Temperature | 42°C |
Reverse Transcriptase | MultiScribe |
Ribonuclease H Activity | Yes |
Includes: MultiScribe™ Reverse Transcriptase (100μL at 50U/μL)
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10117254
|
Applied Biosystems™
N8080234 |
200 Reactions |
£506.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10279624
|
Applied Biosystems™
4304134 |
2,000 Reactions |
£4290.00
1 set |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TaqMan Reverse Transcription Reagents provide the necessary reagents for reverse transcriptase-PCR (RT-PCR). Random hexamers, oligo (dT)16, and reverse primers are included for cDNA synthesis flexibility. These reagents are designed for reverse transcription and polymerase chain reaction amplification of a specific target RNA from either total RNA of mRNA. The reagents in this kit facilitate two-step RT-PCR.
Two-step RT-PCR is performed in two separate reactions, useful when detecting multiple transcripts from a single cDNA reaction, or when storing a portion of the cDNA.
Advantages of TaqMan Reverse Transcription Reagents
• Contains MultiScribe Reverse Transcriptase, a recombinant moloney murine leukemia virus reverse transcriptase (MMLV RT)
• Random hexamers and oligo (dT)16 included for flexibility
• Individual components provide versatility in assay setup
• Compatible with two-step RT-PCR reactions
Using TaqMan Reverse Transcription Reagents
TaqMan Reverse Transcription Reagents provide all the necessary components to perform the reverse transcription of RNA to cDNA. For two-step RT-PCR, random hexamers, oligo (dT)16, and sequence-specific reverse primers can all be used. The choice of primers for reverse transcription is best made after experimentally evaluating all three priming systems.
Specifications
Kit | |
42°C | |
Yes | |
Real Time PCR (qPCR) | |
First-Strand cDNA | |
Reverse Transcription | |
RNA |
Reverse Transcription | |
MultiScribe | |
Dry Ice | |
50 U/μL | |
Separate components | |
Up to 5 kb |
For Research Use Only. Not for use in diagnostic procedures.