Learn More
TaqMan™ TaqMan™ RNase P Instrument Verification Plate, Fast 96-well (for 0.1 mL block)
Satisfies the same requirements as the real-time PCR system installation.
Brand: TaqMan™ 4351979
Includes: Contains one 96-well Fast Reaction Plate preloaded and sealed with TaqMan™ reagents, including RNase P probes and primers.
Description
The TaqMan™ RNase P Fast 96-well Instrument Verification Plate is used to verify the performance of the Applied Biosystems™ 7500 Fast, Applied Biosystems™ 7900 HT Fast, StepOnePlus™, ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex real-time PCR system equipped with a 96-well block. The TaqMan™ RNase P Fast 96-well Instrument Verification Plate is a sealed Optical 96-well Fast Thermal Cycling Plate preloaded with necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye–labeled probe) and template.The TaqMan™ RNase P Fast 96-well Instrument Verification Plate satisfies the same requirements as the real-time PCR system installation. To pass installation, the instruments must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, we recommend that the performance of your instrument be verified every six months.
- Detects and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme
- Satisfies the same requirements as the real-time PCR system installation—the instruments must demonstrate the ability to distinguish between 5000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well
- Performance of instrument should be verified every six months
Order Info
Dry ice shipping; store at -15° to -25°C; protect from light.;
Specifications
Contains one 96-well fast reaction plate for the 0.1 mL block preloaded and sealed with TaqMan™ reagents, including RNase P probes and primers. Store at -15°C to -25°C. Protect from light. |
|
7500 Fast System, 7900HT Fast System, QuantStudio™ 12k Flex, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, StepOnePlus™, Fast Mode, ViiA™ 7 System, QuantStudio™ 6 Pro, QuantStudio™ 7 Pro | |
TaqMan™ | |
Real Time PCR (qPCR) | |
96-well | |
RNase P Calibration, Instrument Verification Plate |
FAM | |
Fluorescence | |
Instrument Verification Plate | |
Dry Ice | |
1 x 96-well plate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2401844 | Certificate of Analysis | 28/01/2025 | 4351979 |
2401842 | Certificate of Analysis | 28/01/2025 | 4351979 |
2401845 | Certificate of Analysis | 28/01/2025 | 4351979 |
2401847 | Certificate of Analysis | 27/01/2025 | 4351979 |
2404858 | Certificate of Analysis | 27/01/2025 | 4351979 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.