Learn More
JULABO™ Aqua Stabil PHMB-free Water Bath Protective Media
Water bath protective media for the market Great Britain.
Brand:  JULABO™ 8940012GB
 View more versions of this product
Product Code. 17857687
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Water bath protective media
  • For the market Great Britain
Specifications
Specifications
12 x 100 mL
20°C to 80°C
None
Water Baths
Protective Media
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Aqua-stabil
Signal Word
  • ||Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Polymeric N,N-Dimethyl-2-hydroxypropylammoniumchloride, Poly(hexamethylenebiguanide) hydrochloride