Learn More
Karl Fischer Aqualine™ Electrolyte A, for Karl Fischer Titration By Coulometry, Fisher Chemical
Anolyte solution for general use in conventional cells with a diaphragm
Brand: Fisher Chemical K/2500/08
34.29 GBP valid until 2025-04-18
Use promo code "24095" to get your promotional price.
Description
- Anolyte solution for use in conventional cells with a diaphragm
- Source of electrolyte required for anodic oxidation for coulometric analysis
- Contains methanol and chloroform as solvents

Specifications
Aqualine™ Electrolyte A | |
Karl Fischer Reagent | |
67-66-3, 108-75-8, 111-42-2, 7446-09-5 | |
Liquid | |
81734 |
Colorless | |
500 mL | |
Amber glass bottle | |
Pass Test | |
1992 |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Aqualine Electrolyte A
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
18Hours
8Minutes
26Seconds
43 -
Multiple Manufacturers
Outlet Corner
Use code 24095 to receive this promotion.Promo Code: 24095Expires: 2025-04-15
Days
3Hours
8Minutes
26Seconds
43 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
49Hours
8Minutes
26Seconds
43New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.