missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Electrolyte CG, for Karl Fischer Titration By Coulometry, Fisher Chemical

Brand:  Fisher Chemical K/2560/04

57.57 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.



Product Code. 10169371

  • £111.00 / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

25mL, Vial

  • Catholyte solution for use in conventional cells with a diaphragm
  • Reagent required to complete the electrochemical reaction in the cathode compartment for coulometric analysis
  • Contains methanol as a solvent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aqualine™ Electrolyte CG
Karl Fischer Reagent
108-75-8, 7446-09-5, 7553-56-2
Vial
Pass Test
Colorless
25 mL
64°C
Liquid
3097388
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2527108 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog NumberK/2560/04CN,K/2560/04
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Fischer Aqualine™ Electrolyte CG, for Karl Fischer Titration By Coulometry, Fisher Chemical > 25mL, Vial

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.