Learn More
Fisherbrand™ 96-Well Polystyrene Plates
98.33 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Ideal for antibiotic screens, cell-based assays and screening compounds
Designed for compatibility with automated equipment.
- Polystyrene
- Plates stack easily without lids
- Available in flat or round well shapes
- Nonsterile
- Untreated, provides uniform surface properties
- Lid is sold separately (12-565-511)
- Cat No 12-565-501 is Clear in color
Specifications
Standard | |
8 x 12 | |
Clear | |
Polystyrene | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
80/Cs. |
Without Lid | |
80 Cases | |
Microplate | |
400 μL | |
50-250 μL/well | |
Non-treated | |
Ideal for antibiotic screens, cell based assays and screening compounds |
Product Content Correction
Product Title
Fisherbrand™ 96-Well Polystyrene Plates > Flat bottom; Clear; Without lid
Spot an opportunity for improvement?Share a Content Correction