missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Aquarium Air Pump

Works with a variety of aquaria with availability in a number of different sizes. Azpack™ Aquarium Air Pump powers aquarium air filters quietly and efficiently.

Brand:  Azpack™

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12473170

  • £78.25 / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

  • Reliable hard wearing pumps with anti-vibration feet
  • Quiet operation
  • Available in different sizes to suit all aquaria
  • Electrical supply: 230V/50Hz
Specifications

Specifications

230 V 50 Hz
2.5 w
1 m
AP2
3.5 L/min.
0.17 mbar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers