missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Aquarium Air Pump
Works with a variety of aquaria with availability in a number of different sizes. Azpack™ Aquarium Air Pump powers aquarium air filters quietly and efficiently.
£67.85 - £169.00
Product Code | Brand | Flow Rate | Maximum Pressure | Model | Overall Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Maximum Pressure | Model | Overall Length (Metric) | Price | Quantity & Availability | ||
12463170
|
Azpack™
12463170 |
2 L/min. | 0.20 mbar | AP1 | 600 mm |
£67.85
Each |
In Stock
Log in to see stock. |
||
12473170
|
Azpack™
12473170 |
3.5 L/min. | 0.17 mbar | AP2 | 1 m |
£78.25
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
12483170
|
Azpack™
12483170 |
6 L/min. | 0.19 mbar | AP3 | 1.2 m |
£109.00
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||
12493170
|
Azpack™
12493170 |
10 L/min. | 0.19 mbar | AP4 | 1.5 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Reliable hard wearing pumps with anti-vibration feet
- Quiet operation
- Available in different sizes to suit all aquaria
- Electrical supply: 230V/50Hz
Specifications
230 V 50 Hz |