missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Aquarium Air Pump

Works with a variety of aquaria with availability in a number of different sizes. Azpack™ Aquarium Air Pump powers aquarium air filters quietly and efficiently.

Brand:  Azpack™

Code : 04

Additional Details : Weight : 0.57240kg

 View more versions of this product

Product Code. 12493170

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Reliable hard wearing pumps with anti-vibration feet
  • Quiet operation
  • Available in different sizes to suit all aquaria
  • Electrical supply: 230V/50Hz
Specifications

Specifications

230 V 50 Hz
7 w
1.5 m
AP4
10 L/min.
0.19 mbar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers