missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Class AS Graduated Pipettes

Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Graduated Pipettes are available in either blue or amber graduation.

Brand:  Hirschmann™ 1100175

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 11762352

  • £137.00 / Pack of 6
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • Acc. to DIN EN ISO 835
  • Main point ring graduation (in either blue or amber)
  • Graduated to tip, zero on tip, type 3
  • With cotton plug end as standard
  • Conformity certified, with dated batch identification
Specifications

Specifications

AS
Yes
0.2 mL
6 Pack
50 mL
Graduated Pipet
AR-GLAS
Amber
Durable amber stain diffusion ink
0.2 mL
450 ±5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers