Learn More
Hirschmann™ Class AS Graduated Pipettes
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Graduated Pipettes are available in either blue or amber graduation.
£70.73 - £137.00
Product Code | Brand | Volume (Metric) | Graduations | Graduation Color | Tolerance | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Graduations | Graduation Color | Tolerance | Price | Quantity & Availability | ||
11682289
|
Hirschmann™
1100133 |
1 mL | 0.01 mL | Amber | 0.007 mL |
£70.75
Pack of 12 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
11767951
|
Hirschmann™
1100160 |
10 mL | 0.1 mL | Amber | 0.05 mL |
£96.45
Pack of 12 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
11720814
|
Hirschmann™
1101129 |
0.5 mL | 0.01 mL | Blue | 0.006 mL |
£89.85
Pack of 12 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
11786612
|
Hirschmann™
1100129 |
0.5 mL | 0.01 mL | Amber | 0.006 mL |
£89.85
Pack of 12 |
In Stock
Log in to see stock. |
||
11740814
|
Hirschmann™
1101135 |
1 mL | 0.1 mL | Blue | 0.007 mL |
£70.73
Pack of 12 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||
11716622
|
Hirschmann™
1100135 |
1 mL | 0.1 mL | Amber | 0.007 mL |
£70.75
Pack of 12 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
11710834
|
Hirschmann™
1101165 |
20 mL | 0.1 mL | Blue | 0.1 mL |
£81.06
Pack of 6 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
11737971
|
Hirschmann™
1100165 |
20 mL | 0.1 mL | Amber | 0.1 mL |
£81.10
Pack of 6 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
15241209
|
Hirschmann™
1101142 |
2 mL | 0.1 mL | Blue | 0.01 mL |
£70.73
Pack of 12 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||
11796612
|
Hirschmann™
1100142 |
2 mL | 0.1 mL | Amber | 0.01 mL |
£70.75
Pack of 12 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
11692289
|
Hirschmann™
1100141 |
2 mL | 0.02 mL | Amber | 0.01 mL |
£70.75
Pack of 12 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
11762352
|
Hirschmann™
1100175 |
50 mL | 0.2 mL | Amber | 0.2 mL |
£137.00
Pack of 6 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||
11757951
|
Hirschmann™
1100153 |
5 mL | 0.05 mL | Amber | 0.03 mL |
£82.65
Pack of 12 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
11790824
|
Hirschmann™
1101154 |
5 mL | 0.1 mL | Blue | 0.03 mL |
£77.96
Pack of 12 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
11706622
|
Hirschmann™
1100154 |
5 mL | 0.1 mL | Amber | 0.03 mL |
£78.00
Pack of 12 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
11747971
|
Hirschmann™
1100170 |
25 mL | 0.1 mL | Amber | 0.1 mL |
£81.10
Pack of 6 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||
11722303
|
Hirschmann™
1100140 |
2 mL | 0.01 mL | Amber | 0.01 mL |
£92.95
Pack of 12 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
Description
- Acc. to DIN EN ISO 835
- Main point ring graduation (in either blue or amber)
- Graduated to tip, zero on tip, type 3
- With cotton plug end as standard
- Conformity certified, with dated batch identification
Specifications
AS | |
AR-GLAS | |
Yes | |
Graduated Pipet |