missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ArC™ Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD™ Fixable dead cell stain kits)

Product Code. 10093324
Click to view available options
No. of Tests:
25
100
Unit Size:
1 set
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10093324

Brand: Invitrogen™ A10346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

ArC™ Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD™ Fixable dead cell stain kits)

ArC™ Amine Reactive Compensation Bead Kit is a bead-based compensation tool specifically optimized for use with LIVE/DEAD™ Fixable dead cell stain kits.

  • Ready-to-use control
  • Eliminates the hassle of heat-treating cells as a control, saving precious sample
  • Enables accurate and consistent results

    View a selection guide for all fixable viability dyes for flow cytometry.

    Ready-to-use control
    Unlike other methods that require sample preparation to make compensation controls using cellular material, the ArC™ Amine Reactive Compensation Bead Kit is ready to use out of the dropper bottle. The kit includes two types of specially modified polystyrene microspheres that have been optimized to react with the amine-reactive dye in the LIVE/DEAD™ Fixable dead cell stain kits. The ArC™ reactive beads (Component A) bind any of the amine-reactive dyes and provide a positive signal; the ArC™ negative beads (Component B) have no reactivity and provide a negative compensation control. This ready-to-use product preserves the original sample material for experiments; precious sample is not used for the creation of compensation controls.

    Accurate and consistent results
    Unlike biological controls, which can have variable expression levels depending upon disease or development state, the Arc™ beads provide a bright, consistent signal (see figure below). Consistency in the staining pattern of compensation controls is important for obtaining the right compensation value. If the signal varies with the control material used, the compensation value can be affected, leading to erroneous results.

    How it works
    One drop of beads is added to a test tube containing LIVE/DEAD™ Fixable dead cell stain. After incubation with amine-reactive dye, the beads are washed in staining buffer, a drop of negative control beads is added (if required), and the beads are resuspended and analyzed by flow cytometry. The two components provide a distinct positive bead that is at least 50-fold brighter than the negative beads, which can be used to set compensation.

    Other compensation products
    Besides the ArC™ Amine Reactive Compensation Beads, Life Technologies also offers the AbC™ Anti-Mouse Bead Kit for capture of mouse anti-human antibodies and AbC™ Anti-Rat/Hamster Bead Kit for capture of rat anti-mouse or hamster anti-mouse antibodies.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Diameter (Metric) 6 μm
    Content And Storage Contains 1 vial ArC™ amine reactive beads and 1 vial of negative beads.
  • Store at 2°C to 8°C
  • Format Tube
    For Use With (Equipment) Flow Cytometer
    No. of Tests 100
    Quantity 1 Kit
    Detection Method Fluorescence
    Excitation Wavelength Range any lasers (UV to 633/635)
    Stain Configuration Surface
    Shipping Condition Room Temperature
    Product Line CompenFlow, Molecular Probes
    Product Type Compensation Beads
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.