missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius arium™ comfort II ASTM 1, 2 Combined Water purification Systems with UV Lamp
Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 2 pure water.
Marque: Sartorius H2O-II-1-UV-T
Description
- Equipped with latest EDI technology and unique cartridge kit
- Innovative iJust technology optimizes product water quality
- Touch display with intuitive menu navigation
- Compact design with SD card slot
- Performs electrochemical deionization in third purification stage
- Reliable and safe removal of contamination contained in feed water
- arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2
- Available as bench-top system for every laboratory workstation and wall-mounted with integrated wall bracket
Benefits
- Optimized, economic water consumption
- Premium product-water quality at all times
- Guaranteed longer life of downstream ultrapure water systems
Spécification
<1 CFU/L | |
5 L/hr. | |
Water Purification System | |
2°C to 30°C | |
80% | |
Benchtop | |
Adsorption using spherical activated carbon, catalyst, reverse osmosis, electrochemical deionization, optional final particle and sterile filtration | |
17 in. | |
19.7 in. | |
18.7 in. | |
61.7 lb. (Empty); 79.3 lb. (Operating) | |
100/240 VAC (+/-10%) 50/60 Hz 130 VA (max.) |
120 L/hr. (Type 1), 5 L/hr. or 10 L/hr. (Type 2) | |
Type 1 and 2 | |
2.0 to 6.9 bar | |
HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC Analysis, Photometry, Microbiological media and reagents, Histology, ELISA, RIA | |
UV lamp | |
<1mL | |
<1 NTU | |
43.5 cm | |
50.1 cm | |
47.6 cm | |
28 kg (Empty); 36 kg (Operating) |