missing translation for 'onlineSavingsMsg'
Learn More

Sartorius arium™ comfort II ASTM 1, 2 Combined Water Purification Systems w/o UV Lamp, TOC Monitor

Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 2 pure water.

Brand:  Sartorius H2O-II-2-B


Code : NEW

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 14851382

  • £13800.00 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Equipped with latest EDI technology and unique cartridge kit
  • Innovative iJust technology optimizes product water quality
  • Touch display with intuitive menu navigation
  • Compact design with SD card slot
  • Performs electrochemical deionization in third purification stage
  • Reliable and safe removal of contamination contained in feed water
  • Arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2
Benefits
  • Optimized, economic water consumption
  • Premium product-water quality at all times
  • Guaranteed longer life of downstream ultrapure water systems
Specifications

Specifications

<1 CFU/L
10 L/hr.
2.0 to 6.9 bar
HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC Analysis, Photometry, Microbiological media and reagents, Histology, LISA, RIA
Wall
Adsorption using spherical activated carbon, catalyst, reverse osmosis, electrochemical deionization, optional final particle and sterile filtration
17 in.
19.7 in.
18.7 in.
61.7 lb. (Empty); 79.3 lb. (Operating)
100/240 VAC (+/-10%) 50/60 Hz 130 VA (max.)
120 L/hr. (Type 1), 5 L/hr. 10 L/hr. (Type 2)
Water Purification System
2°C to 30°C
80%
<1mL
<1 NTU
43.5 cm
50.1 cm
47.6 cm
28 kg (Empty); 36 kg (Operating)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers