missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius arium™ comfort II ASTM 1, 2 Combined Water Purification Systems w/o UV Lamp, TOC Monitor
Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 2 pure water.
£11420.00 - £13800.00
Specifications
Bacterial Content | <1 CFU/L |
---|---|
Daily Output | 120 L/hr. (Type 1), 5 L/hr. 10 L/hr. (Type 2) |
Type | Water Purification System |
Feedwater Pressure | 2.0 to 6.9 bar |
Feedwater Temperature (Metric) | 2°C to 30°C |
Product Code | Brand | Mounting | Flow Rate | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mounting | Flow Rate | Price | Quantity & Availability | ||||
16495354
|
Sartorius
H2O-II-1-T |
Benchtop | 5 L/hr. |
£11420.00
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
14871382
|
Sartorius
H2O-II-2-T |
Benchtop | 10 L/hr. |
£13800.00
Each |
In Stock
Log in to see stock. |
||||
14851382
|
Sartorius
H2O-II-2-B |
Wall | 10 L/hr. |
£13800.00
Each |
In Stock
Log in to see stock. |
||||
Description
- Equipped with latest EDI technology and unique cartridge kit
- Innovative iJust technology optimizes product water quality
- Touch display with intuitive menu navigation
- Compact design with SD card slot
- Performs electrochemical deionization in third purification stage
- Reliable and safe removal of contamination contained in feed water
- Arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2
Benefits
- Optimized, economic water consumption
- Premium product-water quality at all times
- Guaranteed longer life of downstream ultrapure water systems
Specifications
<1 CFU/L | |
Water Purification System | |
2°C to 30°C | |
80% | |
Adsorption using spherical activated carbon, catalyst, reverse osmosis, electrochemical deionization, optional final particle and sterile filtration | |
17 in. | |
19.7 in. | |
18.7 in. | |
61.7 lb. (Empty); 79.3 lb. (Operating) | |
100/240 VAC (+/-10%) 50/60 Hz 130 VA (max.) |
120 L/hr. (Type 1), 5 L/hr. 10 L/hr. (Type 2) | |
2.0 to 6.9 bar | |
HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC Analysis, Photometry, Microbiological media and reagents, Histology, LISA, RIA | |
<1mL | |
<1 NTU | |
43.5 cm | |
50.1 cm | |
47.6 cm | |
28 kg (Empty); 36 kg (Operating) |