missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Arium™ Mini Extend Ultrapure Water System
Compact Arium™ Mini laboratory water systems have been designed for Type 1 ultrapure water requirements of 10 L/day and are ideal for use in preparation of buffers, media and samples, both in life sciences applications and in analytical laboratory procedures.
Brand: Sartorius H2O-MUR-UV-T
This item is not returnable.
View return policy
Description
- Flexible: Different versions for different inlet water and connection options, as well as the desired water withdrawal
- Compact: Space-saving with a width of only 28 cm (+ 9 cm for versions with attached dispensing unit)
- Intuitive: Color touch display, with direct access to all important information and dispensing options
- Innovative: Depending on the type of system, our unique Bagtank technology saves time as no intensive tank cleaning with hazardous substances is required
- Reliable: Delivers consistently high water quality for reliable and reproducible results
Specifications
<0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge ) | |
<0.001 EU/mL with a terminal ultrafiltration cartridge | |
No | |
1 L/min. | |
Optional | |
< 5 ppb | |
Water Purification System |
1 to 10 L | |
3/8 in. | |
Distilled, DI, RO, EDI | |
Type 1 | |
18.2 MΩ x cm | |
≤5 ppb | |
Arium Mini Essential with UV |